missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat IL 12 Protein
A cDNA sequence encoding the IL 12 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP10690-2ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
IL 12 Protein | |
2 μg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Recombinant Protein | |
Baculovirus | |
The protein (1 mg/ml) was lyophilized with no additives. |
Greater than 95.0% as determined by SDS-PAGE. | |
Research Use Only | |
ED50 range = 0.01-0.05 ng/ml corresponding to a specific activity of 20,000,000-100,000,000IU/mg as determined by the dose dependent secretion of IFN-gamma from PHA and IL-2 activated mouse splenocytes that have been costimulated with PMA. Optimal concentration for individual application should be determined by a dose response assay. | |
Rat | |
Untagged |