missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Rat Minpp1 Protein

A DNA sequence encoding the Rattus norvegicus (Rat) Minpp1, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP6360-ye-500ug

Dettagli aggiuntivi : Peso : 0.01000kg

 vedi altre versioni di questo prodotto

Codice prodotto. 16062740

Quantità Prezzo
1 1705.00€ / 500µg
3 1590.94€ / 500µg
Spedizione stimata
21-10-2020
Aggiungi al carrello


Specifica

Specifica

NP_062136.1
Minpp1 Protein
500 μg
Not determined.
Full Length
Rat
His
Tris-based buffer, 50% glycerol
29688
Greater than 90% as determined by SDS-PAGE.
Research Use Only
Minpp1
Recombinant Protein
Yeast
SLPGRGDPVA SVLSPYFGTK TRYEDVNPWL LGDPVAPRRD PELLAGTCTP VQLVALIRHG TRYPTTKQIR KLRQLQGLLQ TRESVDGGSR VAAALDQWPL WYDDWMDGQL VEKGRQDMRQ LALRLAALFP DLFCRENYGR LRLITSSKHR CVDSSAAFLQ GLWQHYHPGL PPPDVSDMEC DPPRVNDKLM RFFDHCEKFL TEVERNATAL YHVEAFKTGP EMQTVLKKVA ATLQVPVNNL NADLIQVAFF TCSFDLAIQG VHSPWCDVFD VDDAKVLEYL NDLKQYWKRS YGYAINSRSS CNLFQDIFLH LDKAVEQKQR SQPVSSSVIL QFGHAETLLP LLSLMGYFKD KEPLTAYNFE EQVHREFRSG HIVPYASNLI FVLYHCEDAQ TPQEKFQIQM LLNEKVLPLA HSQKTVALYE DLKNHYQDIL QSCQTSKECN LPKVNITSDE L
Videos
Documenti