missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat PRNP / Prion Protein
A DNA sequence encoding the Rattus norvegicus (Rat) PRNP / Prion, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP6532-ec-50ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_036763.1 | |
PRNP / Prion Protein | |
Research Use Only | |
Prnp | |
Recombinant Protein | |
E. coli | |
GGWNTGGSRY PGQGSPGGNR YPPQSGGTWG QPHGGGWGQP HGGGWGQPHG GGWGQPHGGG WSQGGGTHNQ WNKPSKPKTN LKHVAGAAAA GAVVGGLGGY MLGSAMSRPM LHFGNDWEDR YYRENMYRYP NQVYYRPVDQ YSNQNNFVHD CVNITIKQHT VTTTTKGENF TETDVKMMER VVEQMCVTQY QKESQAYYDG RRS | |
Greater than 90% as determined by SDS-PAGE. |
24686 | |
50 μg | |
Not determined. | |
Full Length of Mature Protein | |
Rat | |
His | |
Tris-based buffer, 50% glycerol |