missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant S. aureus Replication initiation Protein

A DNA sequence encoding the Staphylococcus aureus Replication initiation protein, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP7349-ec-10ug

Dettagli aggiuntivi : Peso : 0.01000kg

 vedi altre versioni di questo prodotto

Codice prodotto. 16058370

Quantità Prezzo
1 309.00€ / 10µg
6 288.51€ / 10µg
Spedizione stimata
26-10-2020
Aggiungi al carrello


Specifica

Specifica

NP_052693.1
S. aureus Replication initiation Protein
10 μg
Not determined.
Full Length
S. aureus
His-SUMO
Tris-based buffer, 50% glycerol
3355242
Greater than 90% as determined by SDS-PAGE.
Research Use Only
repD
Recombinant Protein
E. coli
MSTENHSNYL QNKDLDNFSK TGYSNSRLSG NFFTTPQPEL SFDAMTIVGN LNKTNAKKLS DFMSTEPQIR LWDILQTKFK AKALQEKVYI EYDKVKADSW DRRNMRVEFN PNKLTHEEML WLKQNIIDYM EDDGFTRLDL AFDFEDDLSD YYAMTDKAVK KTIFYGRNGK PETKYFGVRD SDRFIRIYNK KQERKDNADV EVMSEHLWRV EIELKRDMVD YWNDCFDDLH ILKPDWTTPE KVKEQAMVYL LLNEEGTWGK LERHAKYKYK QLIKEISPID LTELMKSTLK ENEKQLQKQI DFWQREFRFW K
Videos
Documenti