missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Sheep IL12B / P40 Protein
A DNA sequence encoding the Ovis aries (Sheep) IL12B / P40, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP6210-ec-200ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_001009438.1 | |
IL12B / P40 Protein | |
Research Use Only | |
IL12B | |
Recombinant Protein | |
E. coli | |
IWELEKNVYV VELDWYPNAP GETVVLTCDT PEEDGITWTS DQSSEVLGSG KTLTIQVKEF GDAGQYTCHK GGEVLSRSLL LLHKKEDGIW STDILKDQKE PKAKSFLKCE AKDYSGHFTC SWLTAISTNL KFSVKSSRGS SDPRGVTCGA ASLSAEKVSM DHREYNKYTV ECQEGSACPA AEESLPIEVV MEAVHKLKYE NYTSSFFIRD IIKPDPPKNL QLRPLKNSRQ VEVSWEYPDT WSTPHSYFSL TFCVQVQGKN KREKKLFTDQ TSAKVTCHKD ANIRVQARDR YYSSFWSEWA SVSCS | |
Greater than 90% as determined by SDS-PAGE. |
443472 | |
200 μg | |
Not determined. | |
Full Length | |
Sheep | |
His | |
Tris-based buffer, 50% glycerol |