missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Zika Ectodomain Protein
A cDNA sequence encoding the Zika Ectodomain was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP13996-10ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
Zika Ectodomain Protein | |
Research Use Only | |
Recombinant Protein | |
Baculovirus | |
The Zika Ectodomain solution 10mM Sodium phosphate, pH 7.2, 150mM NaCl. |
10 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Zika | |
Untagged | |
Greater than 90.0% as determined by SDS-PAGE. |