Learn More
Soluzione standard Redox
Codice prodotto. 11913093
Cambia vista
Click to view available options
Quantità:
500 ml
Dimensione della confezione:
500mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità unitSize
11913093 500 ml 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Codice prodotto. 11913093 Fornitore Chem-Lab N. del fornitore CL03.1803.0500
Please to purchase this item. Need a web account? Register with us today!
Ensure that electrodes are calibrated correctly for maximum accuracy. Redox Standard Solution, +124mV, Chem-Lab is used with Redox/ORP electrodes with a silver/silver chloride wire in saturated potassium chloride. Millivolt output: 124mV at 25°C. Store away from light.
Redox (reduction–oxidation) reactions cause an electron transfer process that can be measured using a Redox or ORP (oxidation-reduction potential) electrode. The milllivolt (mV) output directly correlates with the redox potential of the reaction. Redox standard solutions help calibrate and verify the measurements of Redox electrodes.
Millivolt Output Specifications:
  • 10°C—141 mV
  • 12°C—138 mV
  • 14°C—136 mV
  • 16°C—134 mV
  • 18°C—132 mV
  • 20°C—130 mV
  • 22°C—127 mV
  • 24°C—125 mV
  • 25°C—124 mV
  • 26°C—123 mV
  • 28°C—125 mV
  • 30°C—122 mV
  • 32°C—120 mV
  • 34°C—117 mV
  • 36°C—115 mV
  • 38°C—112 mV
  • 40°C—99 mV
Specifica
Densità 1.04g/mL
Conducibilità 124mV at 25°C
Da utilizzare con (apparecchiatura) Redox and ORP Electrodes
Quantità 500 ml
Concentration or Composition (by Analyte or Components) K3(Fe(CN)6)
K4(Fe(CN)6) • 3H2O
H2O