Learn More
BRAND™ Reducing Adapter
Polypropylene construction. Simplify connections to most water sources. For use with water jet pump.
Marca: BRAND™ 159665
5.08 EUR valido fino al 2024-04-30
Usa il codice promo "21656" per avere il tuo prezzo promozionale.
Dettagli aggiuntivi : Peso : 0.80000kg
Descrizione
- Polypropylene construction
- Used to simplify connections to most water sources
- For use with water jet pump
Specifica
Water jet filter pump | |
Polypropylene | |
R 3/8 in. | |
Reducing Adapter |