Learn More
Lenz Laborglasintrumente Reduction Adapters with Ground Joints
Descrizione
- Acc. to DIN 12257 and in addition to DIN standard
- Conical ground sockets and cones
- Made of DURAN™ tubing
Specifica
Specifica
| Cone Size | NS 19/26 |
| Materiale | Borosilicate Glass (3.3) |
| N. giunto rastremato standard | Socket NS 34/35 |