missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Reichert™ TS Meter-D Automatic Digital Clinical Refractometer
Automatically read samples with precision comparable to high-end automatic refractometers.
Marca: Reichert™ 13960000
Dettagli aggiuntivi : Peso : 0.80000kg
Descrizione
- New sapphire prism for greater durability and accuracy
- New sapphire prism delivers 2.5x more conductivity to temperature, providing greater accuracy of the Automatic Temperature Compensation (ATC) calculation
- Wide reading range enables use of just one instrument to measure different samples
- Direct reading of urine specific gravity, copper sulfate specific gravity, serum or plasma protein concentration all with great precision
- Touch-of-a-button easy
- No more guessing about fuzzy shadowlines
- PC connectivity for transfer of measurements and programming of scales
- IR Communications Package enables reading log, calibration log, and custom scale transfer to any lab PC
Specifications
- Reading Range: 1.000 to 1.060 Urine Specific Gravity; 0.0 to 15.0g/100mL Serum Protein; 1.000 to 1.200 Copper Sulfate Specific Gravity; 1.3300 to 1.5600 Refractive Index
- Scale: Refractive Index, Urine Specific Gravity, Serum Protein, Copper Sulfate
- Precision: 0.0001 Refractive Index; ±0.001 Urine Specific Gravity; ±0.001 Copper Specific Gravity; ±0.1 g/100mL Serum Protein
- Accuracy: ±0.0001
- Calibration: Set point or set point/span user definable
- Prism Assembly: Sapphire in stainless steel well
- I/O: IR Docking Station
- Power: 4-AAA alkaline batteries (included)
- Dimensions (L x W x H): 18 x 9 x 3.5cm
Servizi
Two years
Specifica
Automatic Digital Clinical Refractometer | |
1.3300 to 1.5600 | |
Set point or set point/span user definable | |
Direct reading of urine specific gravity, copper sulfate specific gravity, serum or plasma protein concentration | |
18 x 9 x 3.5 cm |
1.000 to 1.060 Urine Specific Gravity, 0.0 to 15.0 g/100 mL Serum Protein, 1.000 to 1.200 Copper Sulfate Specific Gravity | |
TS Meter-D | |
±0.0001 | |
NIST | |
4xAAA Alkaline Batteries |