Learn More
rel-(2S,3aR,7aS)-Octahydroindole-2-carboxylic Acid, TRC
Product Code. 30426483
Change view
Click to view available options
Quantità:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantità Unit Size
30426483 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30426483 Supplier Toronto Research Chemicals Supplier No. TRCO2369015MG
 Articolo soggetto a restrizioni
Spedizione stimata: 12-11-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Materiale o nome chimico L-(2S,3aR,7aS)-Octahydroindole-2-carboxylic Acid
CAS 87679-58-1
Formula molecolare C9H15NO2
Quantità 5 mg
InChI Formula InChI=1S/C9H15NO2/c11-9(12)8-5-6-3-1-2-4-7(6)10-8/h6-8,10H,1-5H2,(H,11,12)/t6-,7+,8+/m1/s1
SMILES OC(=O)[C@@H]1C[C@H]2CCCC[C@@H]2N1
Molecular Weight (g/mol) 169.22