Learn More
Thermo Scientific™ Siero agglutinante Remel™, Salmonella H
Codice prodotto. 12789280
Cambia vista
Click to view available options
Descrizione:
Polyvalent, Phase 2
Polyvalent, Phases 1 and 2
Rapid Diagnostic 1 (b, d, E, r)
Rapid Diagnostic 2 (b, E, k, L)
Rapid Diagnostic 3 (d, E, G, k)
Salmonella H (1, 2)
Salmonella H (1, 5)
Salmonella H (1, 6)
Salmonella H (1, 7)
Salmonella H (E) Polyvalent
Salmonella H (L) Polyvalent
Salmonella H (a)
Salmonella H (b)
Salmonella H (c )
Salmonella H (d)
Salmonella H (eh)
Salmonella H (enx)
Salmonella H (gm)
Salmonella H (i)
Salmonella H (r )
Salmonella H (y)
Salmonella H (z27)
Dimensione della confezione:
2mL
Pezzo
22 product options available for selection
Product selection table with 22 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Descrizione Dimensione della confezione
12789280 Salmonella H (d) Pezzo
13245579 Salmonella H (i) 2mL
11933342 Salmonella H (c ) 2mL
11983332 Salmonella H (a) 2mL
12697429 Salmonella H (1, 7) Pezzo
12719290 Salmonella H (enx) Pezzo
11923342 Salmonella H (b) 2mL
12667429 Salmonella H (1, 2) Pezzo
12677429 Salmonella H (1, 5) Pezzo
12687429 Salmonella H (1, 6) Pezzo
12759290 Salmonella H (gm) Pezzo
12677499 Polyvalent, Phases 1 and 2 Pezzo
12759300 Salmonella H (r ) Pezzo
12709290 Salmonella H (eh) Pezzo
12617429 Salmonella H (z27) Pezzo
12749280 Rapid Diagnostic 3 (d, E, G, k) Pezzo
12719300 Salmonella H (L) Polyvalent Pezzo
13235579 Salmonella H (E) Polyvalent Pezzo
12660008 Salmonella H (y) Pezzo
13265579 Polyvalent, Phase 2 2mL
12697579 Rapid Diagnostic 1 (b, d, E, r) Pezzo
12623658 Rapid Diagnostic 2 (b, E, k, L) Pezzo
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 22 options available.
22 options
Codice prodotto. 12789280 Fornitore Thermo Scientific™ N. del fornitore R30160501
  • ONLINE EXCLUSIVE PRICE
    257.00€
    111.07€ / pezzi 1
    Risparmia 145.93€
    56% Sconto
 Articolo soggetto a restrizioni
Spedizione stimata: 15-12-2026
Used for identification of flagellar H antigens of Salmonella strains by slide and/or tube agglutination test.
Remel Agglutinating Sera, Salmonella H is used for identification of flagellar H antigens of Salmonella strains by slide and/or tube agglutination test.
  • Select sera also for use as positive controls for stained Salmonella suspensions
TRUSTED_SUSTAINABILITY
Specifica
Set di controllo Positive
Descrizione Salmonella H (d)
Marchio CE Yes
Analiti rilevabili Antigeni H flagellari di ceppi di Salmonella per test di agglutinazione su vetrino e/o provetta
Tipo Agglutinating Sera
Tipo di imballaggio 2 mL/Vial
Quantità 2 mL
Target Organism Class Salmonella