Learn More
Thermo Scientific™ Remel™ Contact Plate Sterile D/E (Dey-Engley) Neutralizing Agar (Irradiated)
Codice prodotto. 17792638
Cambia vista
Click to view available options
Dimensione della confezione:
pezzi 10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. unitSize
17792638 pezzi 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Codice prodotto. 17792638 Fornitore Thermo Scientific™ N. del fornitore R111803
Please to purchase this item. Need a web account? Register with us today!
Thermo Scientific™ Remel Contact Plate Sterile D/E Neutralizing Agar (Irradiated) is used for the cultivation of microorganisms with neutralization of specific chemical disinfectants.
Thermo Scientific™ Remel Contact Plate Sterile D/E Neutralizing Agar (Irradiated) is used for the cultivation of microorganisms with neutralization of specific chemical disinfectants.
  • Contact Plates Sterile are double-bagged and a third bag is enclosed for customer use (Double + Bag)
  • These products are manufactured in a clean room and validated sterile by gamma radiation
  • The Contact Plates are made using GripFit™ technology, which ensures the lid remains in place during handling and yet can be easily removed during environmental monitoring
TRUSTED_SUSTAINABILITY
Specifica
Descrizione Contact Plates, D/E-Neutralizing Agar
Formato Contact - Double Bag
Product Type Agar Plate
Quantità 10/Pk.
Not intended for IVD use.