Learn More
Thermo Scientific™ Remel™ PathoDx™ Extraction Reagent 3
Descrizione
Remel PathoDx and Extraction Reagent 3 is used with Strep A Latex Agglutination Kit and Strep Grouping Kit.
Specifica
Specifica
| Content And Storage | 2°C to 8°C |
| Descrizione | PathoDx Extraction Reagent 3 |
| Marchio CE | Yes |
| Quantità | Each |