Learn More
Thermo Scientific™ Rotavirus Control Kit
Codice prodotto. 13235599
Cambia vista
Click to view available options
Quantità:
2 Vials/Kit
Dimensione della confezione:
pezzi 2
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità Dimensione della confezione
13235599 2 Vials/Kit pezzi 2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Codice prodotto. 13235599 Fornitore Thermo Scientific™ N. del fornitore R246551
  • ONLINE EXCLUSIVE PRICE
    407.00€
    362.60€ / pezzi 2
    Risparmia 44.40€
    10% Sconto
 Articolo soggetto a restrizioni
Spedizione stimata: 14-01-2027
Monitor gross systematic failures using Xpect Rotavirus Control Kit as external quality control material for Xpect Rotavirus Kit.
Monitor gross systematic failures using Thermo Scientific™ Remel™ Xpect™ Rotavirus Control Kit as external quality control material for Xpect Rotavirus Kit. This kit contains a negative and a positive control.
Monitor gross systematic failures using Xpect Rotavirus Control Kit as external quality control material for Xpect Rotavirus Kit.
  • Ready to Use
  • Used as a confirmatory test before the patient sample is used.
  • Should be tested according to Xpect Rotavirus Instructions For Use (IFU).
This kit is used for qualitative procedures. The test should produce negative and positive results with the corresponding control before testing the patient sample.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
TRUSTED_SUSTAINABILITY
Specifica
Descrizione Control Kit
Include 1mL/Vial positive control; 1mL/Vial negative control
Quantità 2 Vials/Kit