Learn More
Thermo Scientific™ Remel™ Yeast Nitrogen Base w/o Amino Acids
Thermo Scientific™ Remel Yeast Nitrogen Base w/o Amino Acids is used for the identification of yeast based on amino acid and carbohydrate requirements.
Marca:  Thermo Scientific™ R459942
Dettagli aggiuntivi : Peso : 0.50000kg
Codice prodotto. 11993852
  • 315.00€ / 500g
Spedizione stimata: 17-05-2024
per visualizzare lo stock disponibile
Aggiungi al carrello
Descrizione
Descrizione
Thermo Scientific™ Remel Yeast Nitrogen Base w/o Amino Acids is used for the identification of yeast based on amino acid and carbohydrate requirements.
Specifica
Specifica
Yeast Nitrogen Base w/o Amino Acids
Powder
Dehydrated Culture Media
500 g
Offerte speciali
For Laboratory Use Only