Learn More
Wuhan Zhongze Trading Co Ltd™ RephiDuo P Pack S2
Codice prodotto. 15419739 Sfoglia Tutto Fisherbrand Traceable Prodotti
Cambia vista
Click to view available options
Wuhan Zhongze Trading Co Ltd™ RephiDuo P S2
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto.
15419739 Wuhan Zhongze Trading Co Ltd™ RephiDuo P S2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Codice prodotto. 15419739
  • ONLINE EXCLUSIVE PRICE
    381.27€
    309.80€ / pezzi 1
    Risparmia 71.47€
    18% Sconto
Spedizione stimata: 15-02-2027
Aggiungi al carrello
Aggiungi al carrello
RephiDuo P pack S2 for Elix and Rios systems.
RephiDuo P pack S2 for Elix and Rios systems. Comparable to MILPR0G0T0S2.
  • This product is designed to remove large particles and organic compounds, soften water, and remove chlorine from tap water to protect the RO membranes.
Specifica
Tipo Water Purification System