Learn More
Biochrom Resolution CFR Software
Codice prodotto. 17871914
Cambia vista
Click to view available options
Tipo:
Software
Dimensione della confezione:
Pezzo
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Tipo unitSize
17871914 Software Pezzo
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Codice prodotto. 17871914 Fornitore Biochrom N. del fornitore 80710033
Solo null disponibili
Aggiungi al carrello
Aggiungi al carrello
Resolution CFR PC Software for Biochrom spectrophotometers.
  • With password protected user logins and full audit trails, data will be compliant under 21 CFR part 11
  • Quick Read and Quick Scan for fast measurements with limited setup
  • Bespoke method development for your unique laboratory requirements
  • Powerful graphical data display and manipulation options
  • Microsoft look and feel for intuitive use
  • Resolution Life Science version has pre-programmed methods in for fast measurements of commonly methods like protein and DNA/RNA/ quantification and purity measurements as well as cell density measurements and CyDye quantification
  • Library of reference material if your laboratory environment requires regular verification.
  • Save, print or export your analysis as an Excel™, Extended Metafile, HTML, pdf, rich text format, text, Word or XPS file types.
Compatible with Biochrom spectrophotometer models:
  • Novaspec
  • Ultrospec, excludes colorimter and cell density meter
  • WPA, excludes colorimter and cell density meter
  • Libra, excludes colorimter and cell density meter
  • BioDrop
Specifica
Tipo Software