Learn More
Biochrom Resolution CFR Software
Descrizione
- With password protected user logins and full audit trails, data will be compliant under 21 CFR part 11
- Quick Read and Quick Scan for fast measurements with limited setup
- Bespoke method development for your unique laboratory requirements
- Powerful graphical data display and manipulation options
- Microsoft look and feel for intuitive use
- Resolution Life Science version has pre-programmed methods in for fast measurements of commonly methods like protein and DNA/RNA/ quantification and purity measurements as well as cell density measurements and CyDye quantification
- Library of reference material if your laboratory environment requires regular verification.
- Save, print or export your analysis as an Excel™, Extended Metafile, HTML, pdf, rich text format, text, Word or XPS file types.
Compatible with Biochrom spectrophotometer models:
- Novaspec
- Ultrospec, excludes colorimter and cell density meter
- WPA, excludes colorimter and cell density meter
- Libra, excludes colorimter and cell density meter
- BioDrop
Specifica
Specifica
| Tipo | Software |