Learn More
Cytiva RESOURCE™ and SOURCE™ 15 and 5RPC columns
Product Code. 11590804 Shop All Cytiva Products
Change view
Click to view available options
Volume (Metric):
1 mL
3 mL
Unit Size:
1mL
3mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Unit Size
11590804 3 mL 3mL
10249434 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11590804 Supplier Cytiva Supplier No. 17118201
 Restricted Item
In stock
Designed for rapid screening experiments, method development and small-scale protein purification using reversed phase chromatography
  • Wide pH range (1-12), outstanding selectivity, chemical resistance, high capacity and high resolution at high flow rates
  • Excellent scalability from RESOURCE to FineLINE columns
  • Use RESOURCE RPC 3 ml column for higher resolution/method development on a 10 cm bed height
TRUSTED_SUSTAINABILITY
Specifications
Diameter (Metric) 6.4 mm
Flow Rate 10 mL/min
Ligand Type Matrix surface
Max. Pressure 580 psi
Particle Shape Monosized
Particle Size 15 μm
pH 1 to 12
Volume (Metric) 3 mL
Packing Material Polystyrene/Divinyl Benzene
Endcapped No
Material PEEK
Product Type Reversed Phase Column
Show More Show Less
compliance-icons
Product Identifier
  • RESOURCE RPC, 3 ml
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: ethanol
  • UNKNOWN ACUTE-16.5 per cent of the mixture consists of component(s) of unknown acute toxicity.