Learn More
Edmund Bühler GmbH™ Clamp for PVC Rod
Fastens glass vessels to the support base with PVC rod
Marca: Edmund Bühler GmbH™ 0001 138
Dettagli aggiuntivi : Peso : 0.00630kg
Descrizione
Fastens glass vessels to the support base with PVC rodSpecifica
Round bottom vessels | |
NS 29 | |
Distance piece |
32 mm | |
PVC Rods, support base for IRB 6 | |
NS 29 |