Learn More
Brady™ Retractable Barrier Belt with Black Posts
Product Code. 11791146 Shop All Brady Products
Change view
Click to view available options
Color:
Burgundy
Fluorescent Yellow
Green
Length (Metric):
2 m
3.5 m
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Length (Metric) Unit Size
11791146 Fluorescent Yellow 3.5 m Each
11771136 Green 2 m Each
11771146 Burgundy 3.5 m Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11791146 Supplier Brady™ Supplier No. 912671
  • ONLINE EXCLUSIVE PRICE
    765.00€
    688.04€ / pezzi 1
    Risparmia 76.96€
    10% Sconto
Spedizione stimata: 29-12-2026
Retractable Barrier Belt with Black Posts for traffic control.
  • For traffic control.
Specifications
Product Type Stanchion
Alternate Part Number ADAPT BK BARRIER BELT 3,7M 804329 YL FL
Color Fluorescent Yellow
Material Steel
Description Retractable Barrier Belt - 3.5 Meters with Black Posts
For Use With (Application) Traffic Control
Height (English) 3.94 in.
Height (Metric) 100 mm
Includes Retractable Barrier Belt (3.5 m) Black Posts
Length (English) 11.5 ft.
Length (Metric) 3.5 m
Quantity 1 Ea.
Weight (Metric) 12 kg
Width (English) 3.94 in.
Width (Metric) 100 mm