Learn More
RETSCH Knife Mill Grindomix GM300
Codice prodotto. 10574755 Sfoglia Tutto Retsch Prodotti
Cambia vista
Click to view available options
Tensione:
22/230 V
Dimensione della confezione:
Pezzo
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Tensione unitSize
10574755 22/230 V Pezzo
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Codice prodotto. 10574755 Fornitore RETSCH N. del fornitore 20.252.0001
Solo null disponibili
Aggiungi al carrello
Aggiungi al carrello
For size reduction, homogenization and mixing
Instrument for the grinding and homogenization of food stuffs. It allows to process sample volumes up to 4.5L quickly and reproducibly.
  • Four sharp, robust blades and powerful motor which can reach temporary peaks of 3kW
  • Homogenizes substances with high water, oil or fat content as well as for grinding dry, soft and medium-hard products
  • Meets and exceeds all special laboratory and analytical requirements and is a professional device superior to any commercial household mixer
  • Efficient size reduction of up to 4,500mL feed quantity due to a powerful 1.1 kW motor (peak power input >3 kW)
  • Pre- and fine-grinding in one mill: cutting action in regular mode, grinding by impact in reverse mode, pre-grinding in interval mode
  • All parts which come into contact with the sample material are autoclavable
  • Perfect adaptation to application requirements by variable speed from 500 to 4000 min-1 with an increment of 100 min-1
  • Optional gravitation lid for automatic reduction of the grinding chamber volume (Cat. Nos. 08-418-330, -331 and -332)
  • Ten SOPs can be stored
  • Grinding jar in plastic and stainless steel available (Cat. Nos. 08-418-326, -327 and -329)
Specifications
  • Feed material: soft, medium-hard, elastic, containing water/fat/oil, dry, fibrous
  • Size reduction principle: cutting
  • Material feed size†: ã130mm
  • Final fineness†: <300μm
  • Batch size/feed quantity†: 4500mL (w/ standard lid); 4000mL (gravity lid)
  • Grinding chamber volume: 5000mL, reduction with gravity lid
  • Speed setting: digital, 500-4000 min-1
  • Setting of grinding time: digital, 5 sec. to 3 min.
  • Interval operation: yes
  • Interval time: adjustable
  • Storable SOPs: 10
  • Drive: 3-phase asynchronous motor with frequency converter
  • Drive power: continuously 1.1 kW, short term 3 kW
  • Power connection: 1-phase
  • Protection code: IP 20
  • D x W x H (closed): 440 x 440 x 340mm
  • Net weight: ã30kg
Applications:
Agriculture, biology, food, medicine/pharmaceuticals. Examples: candy, cereal bars, cereals, cheese, coated tablets, cocoa nibs, deep-frozen products, dietary supplements, dried and fresh fruit, feed pellets, fish, ham, lettuce, meat, nuts, oil seeds, pharmaceutical materials, plant materials, sausages, soap, spices, vegetables.
† Depending on feed material and instrument configuration/settings.
Specifica
Product Type Knife Mill
Autoclavabile Autoclavable
Capacità (sistema imperiale) 169 oz.
Capacità (sistema metrico) 5000 mL
Certificazione/Conformità CE
Profondità (sistema imperiale) 17.32 in.
Profondità (sistema metrico) 44 cm
Requisiti elettrici 22/230V 50/60 Hz
Da utilizzare con (applicazione) Ideal for agriculture, biology, food, medicine, pharmaceuticals, size reduction, homogenization and mixing
Tempo di macinazione 5 sec. to 3 min.
Altezza (sistema imperiale) 13.38 in.
Altezza (sistema metrico) 34 cm
Hertz 50/60 Hz
Protezione IP IP20
Dimensione particelle <300 μm
Volume campione (sistema imperiale) 152.16 oz.
Volume campione (sistema metrico) 4500 mL
Speed 500 to 4000 min.-1
Tensione 22/230 V
Watt 1100 W
Peso (sistema imperiale) 66.13 lb.
Peso (sistema metrico) 30 kg
Larghezza (sistema imperiale) 17.32 in.
Larghezza (sistema metrico) 44 cm