missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ RHBDD2 Polyclonal Antibody
GREENER_CHOICE

Codice prodotto. 12809012 Sfoglia Tutto Thermo Scientific Prodotti
Cambia vista
Click to view available options
Quantità:
100 μg
Dimensione della confezione:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità unitSize
12809012 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
Codice prodotto. 12809012 Fornitore Invitrogen™ N. del fornitore PA520861

Please to purchase this item. Need a web account? Register with us today!

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

Rabbit Polyclonal Antibody

A suggested positive control is rat lung tissue lysate. PA5-20861 can be used with blocking peptide PEP-0975.

The Rhomboid family of proteins is made up of several widely conserved polytopic membrane serine proteases that play roles in growth and development. Little is known about the role of RHBDD2. Its overexpression in breast carcinoma appears to be an indicator of poor prognosis and may play a role in breast cancer progression. The related proteins RHBDD1 and RHBDD3 are both involved in the regulation of apoptosis, suggesting that RHBDD2 may also be involved in the modulation of apoptotic activity.
TRUSTED_SUSTAINABILITY

Specifica

Antigene RHBDD2
Applicazioni Western Blot, Immunocytochemistry
Classificazione Polyclonal
Concentrazione 1 mg/mL
Coniugato Unconjugated
Formulazione PBS with 0.02% sodium azide
Gene RHBDD2
N. accesso geni Q6NTF9, Q8VEK2
Alias gene 0610011L13Rik; AI480532; H_RG122E10.2a; H_RG122E10.2b; NPD007; Rhbdd2; Rhbdl7; rhomboid domain containing 2; rhomboid domain-containing protein 2; rhomboid, veinlet-like 7; rhomboid-like protein 7; upregulated during skeletal muscle growth 1; Usmg1; WUGSC:H_RG122E10.2a; WUGSC:H_RG122E10.2b
Simboli geni RHBDD2
Specie ospite Rabbit
Immunogeno An 18 amino acid peptide from near the center of human RHBDD2.
Metodo di purificazione Antigen affinity chromatography
Quantità 100 μg
Status giuridico RUO
Primario o secondario Primary
ID gene (immissione) 215160, 360793, 57414
Target Species Human, Mouse, Rat
Content And Storage Maintain refrigerated at 2°C to 8°C for up to 3 months. For long term storage store at -20°C
Product Type Antibody
Forma Liquid
Isotype IgG
Vedi altri risultati Mostra meno risultati
Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.