Learn More
Epredia™ Parapel™ Paraffin Repellent
Product Code. 12582716 Shop All Epredia Products
Change view
Click to view available options
Quantity:
4 oz. pump bottle; 6/CS
Unit Size:
Pack of 6
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
12582716 4 oz. pump bottle; 6/CS Pack of 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12582716 Supplier Epredia™ Supplier No. 2300
  • ONLINE EXCLUSIVE PRICE
    92.35€
    72.06€ / pezzi 6
    Risparmia 20.29€
    21% Sconto
 Restricted Item
In stock
Protect against excess buildup of wax on countertops and instrumentation with Epredia™ Parapel™ Paraffin Repellent.
  • Low odor
  • Reduces daily cleaning times
  • Will not leave behind oily residue
  • Will not remove finish from surfaces
TRUSTED_SUSTAINABILITY
Specifications
Quantity 4 oz. pump bottle; 6/CS
compliance-icons compliance-icons compliance-icons
Product Identifier
  • ParaPel
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 3
  • Acute toxicity Category 1
  • LONG-TERM AQUATIC HAZARD Chronic 2
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H304-May be fatal if swallowed and enters airways.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P331-Do NOT induce vomiting.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.
  • P273-Avoid release to the environment.
  • P370+P378-In case of fire: Use to extinguish.
  • P391-Collect spillage.