Learn More
Epredia™ Richard-Allan Scientific™ Elastic Stain, Van Gieson Stain Solution
Codice prodotto. 12513637 Sfoglia Tutto Epredia Prodotti
Cambia vista
Click to view available options
Product Type:
Alcoholic Hematoxylin Solution
Sodium Thiosulfate Solution
Van Gieson Stain
Weigert's Iodine Solution
Dimensione della confezione:
125mL
250mL
500mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Product Type unitSize
12513637 Alcoholic Hematoxylin Solution 250mL
12543627 Sodium Thiosulfate Solution 500mL
12533637 Weigert's Iodine Solution 125mL
12543637 Van Gieson Stain 250mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Codice prodotto. 12513637 Fornitore Epredia™ N. del fornitore 88007
Please to purchase this item. Need a web account? Register with us today!
Demonstrate the atrophy, thinning and loss of elastic tissue with Epredia™ Richard-Allan Scientific™ Elastic Stain.
  • Based on the Verhoeff Elastic technique
  • Also used for identifying the breaks, splitting and reduplication of elastic lamellae that occur in vascular diseases
  • Elastic fibers are stained blue-black to-black with red collagen and a yellow background
TRUSTED_SUSTAINABILITY
Specifica
Product Type Alcoholic Hematoxylin Solution