Learn More
Epredia™ Signature Series™ Pen-Fix™
Codice prodotto. 13338656 Sfoglia Tutto Thermo Scientific Prodotti
Cambia vista
Click to view available options
Quantità:
5 gal.
4 x 1 gal.
Dimensione della confezione:
Pezzo
pezzi 4
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità Dimensione della confezione
13338656 5 gal. Pezzo
10148438 4 x 1 gal. pezzi 4
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Codice prodotto. 13338656 Fornitore Epredia™ N. del fornitore 6105
  • ONLINE EXCLUSIVE PRICE
    215.14€
    202.00€ / pezzi 1
    Risparmia 13.14€
    6% Sconto
 Articolo soggetto a restrizioni
Spedizione stimata: 22-02-2027
Achieve fixation and dehydration of specimens simultaneously with Epredia™ Signature Series™ Pen-Fix™, a uniquely buffered alcoholic formalin fixative.
  • Excellent frozen section fixative
  • Performs well with fatty or thick tissue sections
  • Penetration rate three times faster than 10% Neutral Buffered Formalin
  • Can be used on all open or enclosed tissue processors and in the grossing room
  • Recommended for use in the second and/or third stations of the processor as a secondary fixative
TRUSTED_SUSTAINABILITY
Specifica
Quantità 5 gal.