Learn More
Epredia™ Richard-Allan Scientific™ Three-Step Stain Kit
Codice prodotto. 10570519 Sfoglia Tutto Epredia Prodotti
Cambia vista
Click to view available options
Product Type:
Epredia Three Step Stain Kit
Solution A
Solution B
Dimensione della confezione:
1 set
Pezzo
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Product Type Dimensione della confezione
10570519 Solution B Pezzo
10530339 Epredia Three Step Stain Kit 1 set
10425339 Solution A Pezzo
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Codice prodotto. 10570519 Fornitore Epredia™ N. del fornitore 3323
  • ONLINE EXCLUSIVE PRICE
    95.20€
    74.31€ / pezzi 1
    Risparmia 20.89€
    21% Sconto
 Articolo soggetto a restrizioni
Spedizione stimata: 23-11-2026
Obtain results on par with Wright-Giemsa with Epredia™ Richard-Allan Scientific™ Three Step Stain.
  • Stain can be used to detect H. Pylori.
  • Set contains easy-to-follow instructions
  • Product is available in one-gallon containers to facilitate use on automated strainers
  • Set contains three 500mL bottles consisting of Fixative Solution, Solution A(Eosin-Y), and Solution B(Azure A and Methylene Blue)
TRUSTED_SUSTAINABILITY
Specifica
Product Type Solution B