Learn More
Rifaximin, TRC
Product Code. 30133298
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
Unit Size:
100mg
1g
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30133298 100 mg 100mg
30012503 1 g 1g
30009790 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30133298 Supplier Toronto Research Chemicals Supplier No. TRCR509500100MG
 Restricted Item
Spedizione stimata: 11-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 80621-81-4
Molecular Formula C43 H51 N3 O11
Molecular Weight (g/mol) 785.88
Synonym (2S,16Z,18E,20S,21S,22R,23R,24R,25S,26R,27S,28E)-5,6,21,23-Tetrahydroxy-27-methoxy-2,4,11,16,20,22,24,26-octamethyl-1,15-dioxo-1,2-dihydro-2,7-(epoxypentadeca[1,11,13]trienoimino)[1]benzofuro[4,5-e]pyrido[1,2-a]benzimidazol-25-yl acetate, 2,7-(Epoxypentadeca[1,11,13]trienimino)benzofuro[4,5-e]pyrido[1,2-a]benzimidazole-1,15(2H)-dione, 25-(acetyloxy)-5,6,21,23-tetrahydroxy-27-methoxy-2,4,11,16,20,22,24,26-octamethyl-, [2S-(2R*,16Z,18E,20R*,21R*,22S*,23S*,24S*,25R*,26S*,27R*,28E)]-, L 105, L 105 (ansamacrolide antibiotic), L 105SV, Rifamycin L 105, Rifamycin L 105SV, Rifaximin, Xifaxan, refaximin
IUPAC Name (2S,16Z,18E,20S,21S,22R,23R,24R,25S,26R,27S,28E)5,6,21,23-Tetrahydroxy-27-methoxy-2,4,11,16,20,22,24,26-octamethyl-1,15-dioxo-1,2-dihydro-2,7-(epoxypentadeca[1,11,13]trienoimino)[1]benzofuro[4,5-e]pyrido[1,2-a]benzimidazol-25-yl acetate
SMILES CO[C@H]1\C=C\O[C@@]2(C)Oc3c(C)c(O)c4c(O)c(NC(=O)\C(=C/C=C/[C@H](C)[C@H](O)[C@@H](C)[C@@H](O)[C@@H](C)[C@H](OC(=O)C)[C@@H]1C)\C)c5c(nc6cc(C)ccn56)c4c3C2=O
Specifications
Quantity 100 mg
InChI Formula InChI=1S/C43H51N3O11/c1-19-14-16-46-28(18-19)44-32-29-30-37(50)25(7)40-31(29)41(52)43(9,57-40)55-17-15-27(54-10)22(4)39(56-26(8)47)24(6)36(49)23(5)35(48)20(2)12-11-13-21(3)42(53)45-33(34(32)46)38(30)51/h11-18,20,22-24,27,35-36,39,48-51H,1-10H3,(H,45,53)/b12-11+,17-15+,21-13-/t20-,22+,23+,24+,27-,35-,36+,39+,43-/m0/s1
Formula Weight 785.3524
Percent Purity >95%
Chemical Name or Material Rifaximin
Grade HPLC