Learn More
Risedronic Acid Sodium Salt, TRC
Product Code. 30132612
Change view
Click to view available options
Quantity:
50 mg
250 mg
2.5 g
Unit Size:
2.5g
250mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30132612 2.5 g 2.5g
30138468 250 mg 250mg
30005752 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30132612 Supplier Toronto Research Chemicals Supplier No. TRCR5215002.5G
 Restricted Item
Spedizione stimata: 10-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 115436-72-1
Molecular Formula C7 H10 N O7 P2 . Na
Molecular Weight (g/mol) 305.09
Synonym Phosphonic acid, P,P'-[1-hydroxy-2-(3-pyridinyl)ethylidene]bis-, sodium salt (1:1), Phosphonic acid, [1-hydroxy-2-(3-pyridinyl)ethylidene]bis-, monosodium salt (9CI), 3-Pyridyl-1-hydroxyethylidene-1,1-biphosphonic acid sodium salt, Actonel, Fossical, Gemfos, NE 58095, Risedronate sodium, Risedronic acid monosodium salt, Sodium risedronate
IUPAC Name sodium;hydroxy-(1-hydroxy-1-phosphono-2-pyridin-3-ylethyl)phosphinate
SMILES [Na+].OC(Cc1cccnc1)(P(=O)(O)O)P(=O)(O)[O-]
Specifications
Quantity 2.5 g
InChI Formula InChI=1S/C7H11NO7P2.Na/c9-7(16(10,11)12,17(13,14)15)4-6-2-1-3-8-5-6;/h1-3,5,9H,4H2,(H2,10,11,12)(H2,13,14,15);/q;+1/p-1
Formula Weight 304.983
Chemical Name or Material Risedronate sodium