missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
RNF36 Polyclonal antibody specifically detects RNF36 in Human,Mouse samples. It is validated for ELISA,Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),Immunocytochemistry/ Immunofluorescence
Specifications
Specifications
| Antigene | RNF36 |
| Applicazioni | ELISA, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence |
| Classificazione | Polyclonal |
| Coniugato | Alexa Fluor 700 |
| Formulazione | 50mM Sodium Borate |
| Alias gene | RFP-like domain-containing protein trimless, RING finger protein 36HSD34, RNF36, Trif, trimless, tripartite motif containing 69, tripartite motif protein 69, tripartite motif-containing 69, tripartite motif-containing protein 69 |
| Specie ospite | Rabbit |
| Immunogeno | Recombinant fusion protein containing a sequence corresponding to amino acids 1-341 of human RNF36 (NP_542783.2).,, Sequence:, MEEELAIQQGQLETTLKELQTLRNMQKEAIAAHKENKLHLQQHVSMEFLKLHQFLHSKEKDILTELREEGKALNEEMELNLSQLQEQCLLAKDMLVSIQAKTEQQNSFDFLKDITTLLHSLEQGMKVLATRELISRKLNLGQYKGPIQYMVWREMQDTLCPGLSPLTLDPKTAHPNLVLSKSQTSVWHGDIKKIMPDDPERFDSSVAVLGSRGFTSGKWYWEVEVAKKTKWTVGVVRESIIRKGSCPLTPEQGFWLLRLRNQTDLKALDLPSFSLTLTNNLDKVGIYLDYEGGQLSFYNAKTMTHIYTFSNTFMEKLYPYFCPCLNDGGENKEPLHILHPQ |
| Metodo di purificazione | Affinity purified |
| Quantità | 0.1 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?