Learn More
Ro-3306, MedChemExpress
Product Code. 16497957
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16497957 5 mg 5mg
16421457 50 mg 50mg
16405248 10 mg 10mg
16435916 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 16497957 Supplier Medchem Express Supplier No. HY12529
 Restricted Item
Spedizione stimata: 09-12-2026
MedChemExpress Ro-3306 is a potent and selective inhibitor of CDK1, with Kis of 20 nM, 35 nM and 340 nM for CDK1, CDK1/cyclin B1 and CDK2/cyclin E, respectively.
Ro-3306 is a potent and selective inhibitor of CDK1, with Kis of 20 nM, 35 nM and 340 nM for CDK1, CDK1/cyclin B1 and CDK2/cyclin E, respectively.
Specifications
Chemical Name or Material Ro-3306
Physical Form Solid
CAS 872573-93-8
Color Brown
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C18H13N3OS2
For Use With (Application) Cancer-Kinase/protease
Solubility Information DMSO : 25 mg/mL (71.13 mM; Need ultrasonic)
SMILES O=C1N=C(NCC2=CC=CS2)S/C1=C\C3=CC=C4N=CC=CC4=C3
Molecular Weight (g/mol) 351.45
Formula Weight 351.45
Percent Purity 98.52%
Grade Research
Show More Show Less
Research purposes only