Descrizione
- The raw materials are free from DiHEMDA und Oleamid. Tips are manufactured at BRAND under monitored cleanroom conditions and are automatically packaged. No parting agents or demolding aids are used. All racked tips and filter tips up to 1000 μL are free of DNA (< 40 fg), RNase (< 8.6 fg), endotoxins (< 1 pg) and ATP (< 1 fg).
- For use with pipetting robots, tips are required that are matched to the robot's mode of operation in terms of geometry and surface properties. BRAND robotic pipette tips and robotic filter tips meet all requirements for use with the Liquid Handling Station pipetting robot.
Specifica
Specifica
| Volume (sistema metrico) | 40 to 1000 μL |
| Filtrato | Con filtro |
| Sterilità | Sterile |
| Imballaggio | TipRack (960/Pk.) |
| Product Type | Robotic Tips |
| Materiale | Polipropilene |