Learn More
Bochem™ Rod for Stand Base
Product Code. 10547122
Change view
Click to view available options
Length (Metric):
1250 mm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Unit Size
10547122 1250 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10547122 Supplier Bochem™ Supplier No. 5154
Top Sellers. Mega Savings.
Use promo code 27997
View all promotions
Spedizione stimata: 22-12-2026
Bochem™ Rod for Stand Base are for use with stand bases.
  • For use with stand bases
Specifications
Material Steel Zincked
Length (Metric) 1250 mm
Diameter (Metric) Outer 12 mm
Thread Size M10
Description External Threads