Learn More
Brady™ Roll Markers
Product Code. 11775573 Shop All Brady Products
Change view
Click to view available options
Display Text:
Air
Eau
Eau incendie
Vapeur
Dimensions (L x W x H):
33 m x 100 mm x 150 mm
33 m x 100 mm x 60 mm
Height (Metric) Character:
8 mm
13 mm
Unit Size:
1 roll
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Display Text Dimensions (L x W x H) Height (Metric) Character Unit Size
11775573 Vapeur 33 m x 100 mm x 60 mm 8 mm 1 roll
11755573 Air 33 m x 100 mm x 150 mm 13 mm 1 roll
11745573 Air 33 m x 100 mm x 60 mm 8 mm 1 roll
11765573 Eau 33 m x 100 mm x 60 mm 8 mm 1 roll
11735573 Eau incendie 33 m x 100 mm x 60 mm 8 mm 1 roll
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 11775573 Supplier Brady™ Supplier No. 226758
  • ONLINE EXCLUSIVE PRICE
    340.00€
    306.67€ / 1 rotoli
    Risparmia 33.33€
    9% Sconto
Spedizione stimata: 07-12-2026
Specifications
Certifications/Compliance European Directives 67/548 and 88/379
Adhesive Type Permanent Acrylic
Color Black on Gray
Format Roll
Material Polyester
Product Type Pipe Marker
Display Text Vapeur
For Use With (Application) Pipe and valve marking
Languages French
No. per Pack 505 Markers/Roll
Quantity 505 Pk.
Temperature Range (English) -40°F to +284°F
Temperature Range (Metric) -40°C to +140°C
Thickness (English) 0.0047 in.
Thickness (Metric) 0.119 mm
Dimensions (L x W) 33 m x 100 mm
Dimensions (L x W x H) 33 m x 100 mm x 60 mm
Length (English) 108.27 ft.
Length (Metric) 33 m
Width (English) 3.94 in.
Width (Metric) 100 mm
Height (English) 2.36 in.
Height (English) Character 0.31 in.
Height (Metric) 60 mm
Height (Metric) Character 8 mm
Alternate Part Number PMFL-VAPEUR-GY3-RLL090