Learn More
Rosmarinic acid, 97+%
Codice prodotto. 16319495
Cambia vista
Click to view available options
Quantità:
1 g
Dimensione della confezione:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità unitSize
16319495 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Codice prodotto. 16319495 Fornitore Thermo Scientific Alfa Aesar N. del fornitore H37451.03
Richiesta di formato massivo o personalizzato
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Specifica
Quantità 1 g