Learn More
Heidolph™ Rotavac Vario Pumping Units
Sfoglia Tutto Heidolph Instruments Prodotti
Click to view available options
Tipo di pompa:
Pumping Unit
Dimensione della confezione:
Pezzo
Descrizione
- Three-stage diaphragm pump is made from chemical-resistant material
- High suction capacity of 1.7 m3/h for fastest evacuation
- Achieves an ultimate vacuum of 5mbar
- Precise vacuum control avoids bumping and eliminates foaming of your evaporation solution
- Can be combined with a condenser
Specifica
Specifica
| Dimensioni (L x P x A) | 263 x 193 x 299 mm |
| Peso (sistema metrico) | 6 kg |
| Tipo di pompa | Pumping Unit |
| Valori nominali armadio | IP65 |