Learn More
Heidolph™ Rotavac Vario Tec RPM-Regulated Pump
Sfoglia Tutto Heidolph Instruments Prodotti
Click to view available options
Maximum Pressure:
12 Mbar
Dimensione della confezione:
Pezzo
Descrizione
- Two-stage diaphragm pump is made from chemical-resistant material.
- Recommended for solvents with low or medium boiling points.
- Can be combined with a condenser.
- Speed adjustment stops completely if vacuum is reached and runs intermittently at very low speed to hold vacuum constant.
- Increased diaphragm performance life due to periods of non-operation.
- Low energy consumption due to intermittent operation.
- Significant noise and vibration reduction due to periods of non-operation.
- Operates without vacuum valve.
- Precise vacuum level at all times.
Specifica
Specifica
| Dimensioni (L x P x A) | 156 x 236 x 196 mm |
| Descrizione | Rotary evaporator vacuum pump |
| Wattage | 160 W |
| Peso (sistema metrico) | 4.3 kg |
| Valori nominali armadio | IP65 |
| Maximum Pressure | 12 Mbar |