Learn More
IKA™ ROTAVISC PT100.8 Temperature Sensor
Product Code. 17247539 Shop All IKA Products
Change view
Click to view available options
For Use With (Equipment):
IKA™ ROTAVISC Viscometer
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
17247539 IKA™ ROTAVISC Viscometer Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17247539 Supplier IKA™ Supplier No. 0025000413
  • ONLINE EXCLUSIVE PRICE
    73.00€
    63.37€ / pezzi 1
    Risparmia 9.63€
    13% Sconto
Spedizione stimata: 28-01-2027
The ROTAVISC PT100.8 Temperature Sensor for use with IKA™ ROTAVISC Rotational Viscometers.
  • Temperature sensor for IKA™ ROTAVISC Rotational Viscometers
  • Durable stainless steel design
  • 3 mm diameter
TRUSTED_SUSTAINABILITY
Specifications
Product Type Temperature Probe (Immersion)
For Use With (Equipment) IKA™ ROTAVISC Viscometer
Material AISI 316Ti
Temperature Range (Metric) -20°C to +350°C