Learn More
Descripción
- Instead of a jewel bearing, these devices are equipped with a special ball bearing technology.
- They are characterized by their high robustness. Predestined for applications with very high sample throughput, frequent spindle changes, operation in harsher environments, or even frequently changing device locations.
- Whether simple or demanding viscosity measurements - with ROTAVISC you get fast and accurate results.
- Viscosity measurement in motion
- The motorized IKA HELISTAND moves the mounted viscometer and its rotating body slowly and uniformly up and down in the sample
- Easy to use
- Intuitive menu navigation awaits you via the generous 4.3'' TFT display. A digital leveling also supports the correct setting of the starting condition. Recurring tasks are facilitated by the simple ramp function.
- Highest measurement accuracy
- The accuracy of the ROTAVISC rotary viscometers for Newtonian and non-Newtonian liquids is ± 1 % of the measuring range. The reproducibility is ± 0.2%.
- Infinitely variable speed
- With ROTAVISC you have a viscometer in which you can adjust the speed stepless, but also specify programs or individual speed levels.
- IKA Rotavisc SBS me-vi HELI Complete
- Viscosity measuring range: 100 to 40,000,000 mPas
Especificaciones
Especificaciones
| Product Type | Viscometer |
| Intervallo viscosità | 100 to 40,000,000 mPa |
| Tipo di display | TFT |
| Accuracy | 0.01 |
| Ripetibilità | 0.002 |
| Tipo di sonda | PT 100 |
| Intervallo di velocità | 0.01 to 200 rpm |
| Temperature Range (Metric) | -100°C to +300°C |
| Risoluzione del termometro | 0.1 K |
| Intervallo coppia | 0.7187 mNm |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?