Description
- Max. speed: 21,300 x g (15,060 rpm)
- Max. capacity: 24 x 1.5/2.0 mL tubes
- Aerosol-tight with Eppendorf QuickLock™ Pro lid
Specifications
Specifications
| Materiale | Aluminum |
| Tipo | Fixed Angle Rotor with Aerosol-tight Lid |
| Capacità | 24 x 1.5/2.0 mL Tubes |
| RCF max | 21,300 x g |
| Velocità max | 15,060 rpm |
| Da utilizzare con (apparecchiatura) | Centrifuge 5425/5425R |
| Certificazione/Conformità | IVD |
| Include | Rotor lid with QuickLock Pro |
| No. of Places | 24 |
| Biocontainment | Aerosol-tight |
| Show More |