Description
- Precision-engineered for optimal sample separation
- All fixed-angle rotors, rotor lids, buckets, caps and adapters can be autoclaved (121°C, 20 min.)
- Accessories (adaptors and buckets) further customize your centrifuge setup
- Aerosol-tight options available
Specifications
Specifications
| Descrizione | FA-24x2, Aerosol-tight - Includes rotor lid |
| Materiale | Anodized Aluminum |
| Tipo | Rotor |
| Capacità | 24 x 1.5/2.0 mL |
| RCF max | 21,100 x g |
| Velocità max | 14,800 rpm |
| Da utilizzare con (apparecchiatura) | Centrifuge 5420 |
| Include | Rotor FA-24x2 for Centrifuge 5420 |