Learn More
Glassco Laboratory Equipments Round Bottom Glass Flask
Product Code. 31937695
Change view
Click to view available options
Capacity (Metric):
25 mL
50 mL
100 mL
250 mL
500 mL
1000 mL
2000 mL
Standard Taper Joint:
14/23
29/32
Unit Size:
Pack of 10
Pack of 6
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Standard Taper Joint Unit Size
31937695 100 mL 29/32 Pack of 10
31937697 25 mL 14/23 Pack of 10
31937677 50 mL 14/23 Pack of 10
31937659 50 mL 29/32 Pack of 10
31937692 100 mL 14/23 Pack of 10
31937676 250 mL 29/32 Pack of 10
31937688 500 mL 29/32 Pack of 10
31937654 1000 mL 29/32 Pack of 10
31937669 2000 mL 29/32 Pack of 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Product Code. 31937695 Supplier Glassco Laboratory Equipments Supplier No. EU057.202.14
Spedizione stimata: 16-12-2026
Add to basket
Add to basket
Round bottom glass flask with a single neck and joint for laboratory use.
  • Round bottom glass flasks with a single neck and joint
  • Available in 25 mL, 50 mL, 100 mL, 250 mL, 500 mL, 1000 mL, and 2000 mL capacities
  • NS 14/23 joints are available for 25 mL, 50 mL, and 100 mL flasks
  • NS 29/32 joints are available for 50 mL, 100 mL, 250 mL, 500 mL, 1000 mL, and 2000 mL flasks
  • Made from heat-resistant ASTM E-438 Type 1, Boro 3.3 glass
  • Complies with DIN/ISO 4797 and USP standards
  • Features white printing
  • Chemical resistant
Specifications
Capacity (Metric) 100 mL
Material Borosilicate Glass (3.3)
Product Type Flask
Standard Taper Joint 29/32
Autoclavable Autoclavable
Disposable Reusable
Graduated No
Mouth Style Joint
Sterility Non-sterile
Bottom Shape Round