Learn More
Eppendorf™ Round Buckets for S-4x400 Rotor
Product Code. 15853212 Shop All Eppendorf Products
Change view
Click to view available options
Quantity:
2 Pk.
4 Pk.
Unit Size:
Pack of 2
Pack of 4
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15853212 2 Pk. Pack of 2
15843212 4 Pk. Pack of 4
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15853212 Supplier Eppendorf™ Supplier No. 5895183000
In stock
These Round Buckets for S-4x400 Rotor are suitable swing-buckets for the S-4x400 rotor that feature a round shape and a 400 mL capacity.
  • Suitable for S-4x400 rotor
  • Round shape
  • Swing-out bucket
  • Variable places depending on adapter used
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Rotor S-4x400
Capacity 400 mL
Quantity 2 Pk.