Learn More
W.S. TYLER™ Round Sieve Brush
Product Code. 10073543 Shop All Cole-Parmer Products
Change view
Click to view available options
For Use With (Equipment):
Test Sieves
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
10073543 Test Sieves Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10073543 Supplier W.S. TYLER™ Supplier No. 5998696
  • ONLINE EXCLUSIVE PRICE
    57.60€
    40.46€ / pezzi 1
    Risparmia 17.14€
    29% Sconto
Spedizione stimata: 18-12-2026
W.S. Tyler round sieve brushes are ideal for cleaning fine and coarse mesh sieves, 120 mesh and finer. Proper maintenance and cleaning of sieves prevents damage to sieve openings, keeping them clean and free of particles, and ensures the longest possible in-spec service life and accurate results.
  • For 120 mesh sieves, and finer
  • Keeps sieves clean for better separation
  • Will not damage or hinder sieve performance
  • Nylon bristles
Specifications
Material Nylon Bristle, Wooden Handle
Description Sieve Cleaning Brush
For Use With (Application) Sieve Cleaning
For Use With (Equipment) Test Sieves
Particle Size Clean 120 Mesh and Finer