Learn More
Roxatidine Acetate Hydrochloride, TRC
Product Code. 30416084
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30416084 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30416084 Supplier Toronto Research Chemicals Supplier No. TRCR7008705G
 Restricted Item
Spedizione stimata: 11-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Roxatidine Acetate Hydrochloride
CAS 93793-83-0
Molecular Formula C19 H28 N2 O4 . Cl H
Quantity 5 g
Synonym Acetamide','2-(acetyloxy)-N-[3-[3-(1-piperidinylmethyl)phenoxy]propyl]-','hydrochloride (1:1), Acetamide','2-(acetyloxy)-N-[3-[3-(1-piperidinylmethyl)phenoxy]propyl]-','monohydrochloride (9CI), Altat, Gastralgin, HOE 760, Neo H2, Roxatidine acetate hydrochloride, Roxit, TZU 0460, Zarocs
InChI Formula InChI=1S/C19H28N2O4.ClH/c1-16(22)25-15-19(23)20-9-6-12-24-18-8-5-7-17(13-18)14-21-10-3-2-4-11-21;/h5,7-8,13H,2-4,6,9-12,14-15H2,1H3,(H,20,23);1H
SMILES Cl.CC(=O)OCC(=O)NCCCOc1cccc(CN2CCCCC2)c1
IUPAC Name [2-oxo-2-[3-[3-(piperidin-1-ylmethyl)phenoxy]propylamino]ethyl] acetate;hydrochloride
Molecular Weight (g/mol) 384.9
Formula Weight 384.1816