missing translation for 'onlineSavingsMsg'
Learn More

RPL27 Mouse anti-Human, Mouse, Clone: OTI6D3, lyophilized, TrueMAB™

Codice prodotto. 17631452
Encompass_Preferred
Cambia vista
Click to view available options
Quantità:
100 μg
Dimensione della confezione:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità unitSize
17631452 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
Codice prodotto. 17631452 Fornitore Origene Technologies N. del fornitore CF810903

Please to purchase this item. Need a web account? Register with us today!

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

Mouse Monoclonal Antibody

Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO

Ribosomes, the organelles that catalyze protein synthesis, consist of a small 40S subunit and a large 60S subunit. Together these subunits are composed of 4 RNA species and approximately 80 structurally distinct proteins. This gene encodes a ribosomal protein that is a component of the 60S subunit. The protein belongs to the L27E family of ribosomal proteins. It is located in the cytoplasm. As is typical for genes encoding ribosomal proteins, there are multiple processed pseudogenes of this gene dispersed through the genome.Ribosomes, the organelles that catalyze protein synthesis, consist of a small 40S subunit and a large 60S subunit. Together these subunits are composed of 4 RNA species and approximately 80 structurally distinct proteins. This gene encodes a ribosomal protein that is a component of the 60S subunit. The protein belongs to the L27E family of ribosomal proteins. It is located in the cytoplasm. As is typical for genes encoding ribosomal proteins, there are multiple processed pseudogenes of this gene dispersed through the genome.

Specifica

Antigene RPL27
Applicazioni Immunohistochemistry (Paraffin), Western Blot
Classificazione Monoclonal
Clone OTI6D3
Coniugato Unconjugated
Formulazione PBS with 8% trehalose and no preservative; pH 7.3
Gene RPL27
N. accesso geni P61353, P61358
Alias gene DBA16, L27
Simboli geni RPL27
Specie ospite Mouse
Immunogeno Full length human recombit protein of human RPL27 produced in E.coli.
Metodo di purificazione Affinity Chromatography
Quantità 100 μg
Status giuridico RUO
Primario o secondario Primary
ID gene (immissione) 19942, 6155
Target Species Human, Mouse
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Forma Lyophilized
Isotype IgG1
Vedi altri risultati Mostra meno risultati
Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.