Learn More
R&D Systems™ RS 246204
Descrizione
RS 246204 is a R-Spondin-1 substitute. Able to initiate formation and growth of mouse intestinal organoids in media without R-Spondin-1 protein. In vivo, RS 24620 has regenerative effect in damaged intestinal epithelium mouse model.
Specifica
Specifica
| Materiale o nome chimico | 3,7-Dihydro-3,7-dimethyl-8-(2-quinolinylthio)-1H-purine-2,6-dione |
| CAS | 878451-87-7 |
| Quantità | 50 mg |
| Linea di prodotti | Tocris |
| Bersaglio | Compounds for Stem Cell Proliferation & Renewal |
| Formula molecolare | C16H13N5O2S |
| Product Type | Bioactive Small Molecule |
| Purezza | 98% |