Learn More
Brady™ Rugged Canvas Lockout Bag
Product Code. 11721925 Shop All Brady Products
Change view
Click to view available options
Color:
Black on Yellow
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
11721925 Black on Yellow Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11721925 Supplier Brady™ Supplier No. 065780
  • ONLINE EXCLUSIVE PRICE
    46.10€
    42.31€ / pezzi 1
    Risparmia 3.79€
    8% Sconto
In stock
This highly visible yellow Lockout Bag is made of rugged canvas with a drawstring and can be locked closed. For securing plugs on portable appliances, cables on pneumatic connectors, crane pendants, etc. Contents not included.
  • Contents not included.
Specifications
Product Type Lockout Bag
Alternate Part Number L/O BAG YELLOW CANVAS DUCK BAG
Color Black on Yellow
Material PVC
Description Lockout bag made of rugged canvas with drawstrings
Dimensions (L x W) 247.65 x 457.20 mm
Quantity 1 Ea.