Learn More
MedChemExpress Rusalatide acetate
Specifica
Specifica
| Purezza | 98.26% |
| CAS | 875455-82-6 |
| Content And Storage | Sealed storage, away from moisture and light∣Powder -80°C 2 years, -20°C 1 year∣In solvent : -80°C, 6 months∣-20°C, 1 month (sealed storage, away from moisture and light) |
| Da utilizzare con (applicazione) | COVID-19-immunoregulation |
| Peso molecolare | 2371.5 g/mol |
| Product Type | Peptides |
| Quantità | 1 mg |
| Shipping Condition | Room temperature |
| Sinonimo | TP508 amide acetate |