missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RXFP4 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen PA549735
Dettagli aggiuntivi : Peso : 0.01000kg
Descrizione
The antibody detects endogenous level of total RXFP4 / GPR100 protein.
RXFP4 Polyclonal antibody specifically detects RXFP4 in Human, Mouse samples. It is validated for ELISA, Western Blot.Specifica
RXFP4 | |
Polyclonal | |
Unconjugated | |
RXFP4 | |
G protein-coupled receptor 100; GPCR142; GPR100; G-protein coupled receptor 100; G-protein coupled receptor GPCR142; Insulin-like peptide INSL5 receptor; JEG62; MGC126556; MGC126558; relaxin 3 receptor 2; relaxin family peptide receptor 4; relaxin/insulin like family peptide receptor 4; relaxin/insulin-like family peptide receptor 4; Relaxin-3 receptor 2; relaxin-3/INSL7 receptor 2; RLN3 receptor 2; Rln3r2; Rxfp4; RXFPR4; SALPR | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
242093, 339403 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
2.30 mg/mL | |
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
Q7TQP4, Q8TDU9 | |
RXFP4 | |
Synthetic peptide corresponding to a region derived from 360-374 amino acids of human elaxin/insulin-like family peptide receptor 4 | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |